missing translation for 'onlineSavingsMsg'
Learn More

Potassium Metabisulfite (2:1), TRC

Product Code. 30012176
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30012176 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30012176 Supplier Toronto Research Chemicals Supplier No. TRCP69807010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 16731-55-8
Quantity 10 g
Molecular Formula K2O5S2
Synonym Disulfurous Acid Dipotassium Salt, Potassium Pyrosulfite, Pyrosulfurous Acid Dipotassium Salt, Dipotassium Disulfite, Dipotassium Metabisulfite, Dipotassium Pyrosulfite, E 224, Kadifit, Potassium Disulfite (K2S2O5), Potassium Metabisulfite, Potassium Metabisulfite (K2S2O5), Potassium Pyrosulfite (K2S2O5)
InChI Formula InChI=1S/2K.H2O5S2/c;;1-6(2)7(3,4)5/h;;(H,1,2)(H,3,4,5)/q2*+1;/p-2
SMILES O=S(S(=O)(O[K])=O)O[K]
Molecular Weight (g/mol) 222.32
Formula Weight 221.8461
Chemical Name or Material Potassium Metabisulfite (2:1)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.