missing translation for 'onlineSavingsMsg'
Learn More

Potassium (meta)Periodate, ≥99.8% (dried material, RT), Honeywell Fluka™

Product Code. 15615980 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15615980 500 g Glass bottle 500g
15605980 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15615980 Supplier Honeywell-Fluka Supplier No. 60450500G
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Puriss. p.a., ACS Reagent,≥99.8% (dried material, RT)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7790-21-8
Molecular Formula IKO4
Molecular Weight (g/mol) 229.999
MDL Number MFCD00011407
InChI Key FJVZDOGVDJCCCR-UHFFFAOYSA-M
Synonym potassium periodate, potassium metaperiodate, unii-q3loc6p66l, q3loc6p66l, periodic acid hio4 , potassium salt, potassium ion io4, ssmicroaea 1/4 o, acmc-20ajue, ssmicroaea 1/4 oni, periodic acid hio4 , potassium salt 1:1
PubChem CID 516896
IUPAC Name potassium;periodate
SMILES [O-]I(=O)(=O)=O.[K+]

Specifications

Quantity 500 g
Linear Formula KIO4
UN Number UN1479
Formula Weight 230g/mol
Percent Purity ≥99.8% (dried material, RT)
Packaging Glass bottle
Chemical Name or Material Potassium (meta)periodate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.