missing translation for 'onlineSavingsMsg'
Learn More

Potassium Iodide, Certified AR for Analysis, Fisher Chemical™

Product Code. p-7073436 Shop All Fisher Chemical Products
Click to view available options
Quantity:
100 g
250 g
500 g
3 kg
Unit Size:
100g
250g
3kg
500g
This item is not returnable. View return policy
For a copy of the TSE/BSE statement please contact Technical Support on Tel: 01509 555888 or email our support team for further assistance
 

Product Code. 10607232

Brand: Fisher Chemical P/5880/48

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For a copy of the TSE/BSE statement please contact Technical Support on Tel: 01509 555888 or email our support team for further assistance
 

IK, CAS Number-7681-11-0, kali iodide, asmofug e, thyro-block, pima, potassium monoiodide, knollide, kisol, potassium diiodide, potassium iodide ki, potassium iodide, 500g, 1330 deg.C, CHEBI:8346, White, 166g/mol, NLKNQRATVPKPDG-UHFFFAOYSA-M, potassium;iodide, 680 deg.C, 11405, 166.003

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7681-11-0
Molecular Formula IK
Molecular Weight (g/mol) 166.003
MDL Number 11405
InChI Key NLKNQRATVPKPDG-UHFFFAOYSA-M
Synonym potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide
PubChem CID 4875
ChEBI CHEBI:8346
IUPAC Name potassium;iodide
SMILES [K+].[I-]

Specifications

Boiling Point 1330°C
Melting Point 680°C
Color White
pH 6 to 8
Physical Form Solid
Quantity 100 g
Formula Weight 166g/mol
Packaging Plastic powder jar
Vapor Pressure 1mmHg 745
compliance-icons
Product Identifier
  • Potassium iodide
Signal Word
  • Danger
Hazard Category
  • Specific target organ toxicity Category 1
Hazard Statement
  • H372-Causes damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P314-Get medical advice/attention if you feel unwell.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.