missing translation for 'onlineSavingsMsg'
Learn More

Potassium iodide, ACS Reagent, ≥99.0%, Solstice

Product Code. 16645763
Change view
Click to view available options
25 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
16645763 25 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16645763 Supplier Fluka Supplier No. 22194525KG
  • ONLINE EXCLUSIVE PRICE
    £7740.00
    £7132.78 / 25kg
    Save £607.22
    7% Off
 Restricted Item
Request Bulk or Custom Formats
Currently Unavailable: We have no information when this product will be available.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity and accuracy assured

TRUSTED_SUSTAINABILITY

Specifications

CAS 7681-11-0
Quantity 25 kg
Molecular Formula IK
InChI Key NLKNQRATVPKPDG-UHFFFAOYSA-M
SMILES [K+].[I-]
IUPAC Name potassium iodide
Molecular Weight (g/mol) 166.00
Chemical Name or Material Potassium iodide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.