missing translation for 'onlineSavingsMsg'
Learn More

Potassium iodide, 99+%, ACS reagent

Product Code. 10297373 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
500 g
1 kg
5 kg
Unit Size:
1kg
500g
5kg
This item is not returnable. View return policy

Product Code. 10297373

Brand: Thermo Scientific Acros 418260050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7681-11-0 | IK | 166 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7681-11-0
Molecular Formula IK
MDL Number MFCD00011405
InChI Key NLKNQRATVPKPDG-UHFFFAOYSA-M
Synonym potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide
PubChem CID 4875
ChEBI CHEBI:8346
IUPAC Name potassium;iodide
SMILES [K+].[I-]

Specifications

Boiling Point 1330.0°C
Melting Point 680.0°C
Color White
pH 6.0 to 9.2 (5% soln. at 25°C)
Physical Form Crystals or Granules
Quantity 5 kg
Assay Percent Range 99+%
Linear Formula KI
Fieser 06,227; 11,440
Merck Index 15, 7764
Solubility Information Solubility in water: 1.43 kg/l. Other solubilities: 45g/L ethanol
Molecular Weight (g/mol) 166
Formula Weight 166
Percent Purity ≥99%
Grade ACS Reagent
Loss on Drying 0.2% max. (150°C, 6 hrs)
Packaging Glass Bottle
Vapor Pressure 1mmHg at 745°C
Density 3.13
Chemical Name or Material Potassium iodide, granular
Show More Show Less
compliance-icons
Product Identifier
  • Potassium iodide
Signal Word
  • Danger
Hazard Category
  • Specific target organ toxicity Category 1
Hazard Statement
  • H372-Causes damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P314-Get medical advice/attention if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.