missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium iodate concentrate, For 1L standard solution, 1/60 M KIO3 (0.1N), Honeywell Fluka™
Click to view available options
Quantity:
1 Pc.
6 x 1 Ea.
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Quantity | 1 Pc. |
Linear Formula | KIO3 |
UN Number | NONH for all modes of transport |
Formula Weight | 214g/mol |
Packaging | Ampoule |
Chemical Name or Material | Potassium iodate concentrate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'