missing translation for 'onlineSavingsMsg'
Learn More

Potassium iodate concentrate, For 1L standard solution, 1/60 M KIO3 (0.1N), Solstice

Product Code. 15695220 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
1 Pc.
6 x 1 Ea.
Unit Size:
Each
Pack of 6
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15695220 6 x 1 Ea. Pack of 6
15685220 1 Pc. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15695220 Supplier Honeywell-Fluka Supplier No. 381206X1EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

for 1L standard solution, 1/60 M KIO3 (0.1N)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula IKO3
Molecular Weight (g/mol) 214.00
MDL Number MFCD00011406
InChI Key JLKDVMWYMMLWTI-UHFFFAOYSA-M
Synonym potassium iodate, potassium triodate, caswell no. 693a, iodic acid, potassium salt, unii-i139e44nhl, potassium iodine oxide kio3, iodic acid hio3 , potassium salt, epa pesticide chemical code 075703, iodic acid hio3 , potassium salt 1:1, kaliumjodat
PubChem CID 23665710
IUPAC Name potassium;iodate
SMILES [O-]I(=O)=O.[K+]

Specifications

Boiling Point 100°C
CAS 7758-05-6, 7732-18-5
CAS Min % >3.0000%, >95.0000%
CAS Max % <5.0000%, <97.0000%
Physical Form Concentrate
Quantity 6 x 1 Ea.
Linear Formula KIO3
UN Number NONH for all modes of transport
Formula Weight 214.00
Packaging Ampoule
Chemical Name or Material Potassium iodate concentrate, For 1L standard solution, 1/60 M KIO3 (0.1N)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.