missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium iodate, 0.1N Standardized Solution, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 L
4 L
Description
Potassium iodate 0.1N solution is used as a standard solution in volumetric titrations. It is extensively used in analytical chemistry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Colorless |
Concentration | 0.1N |
Solubility Information | Miscible with water,potassium iodide solution and sulfuric acid. |
Formula Weight | 214 |
Odor | Odorless |
Quantity | 1 L |
Physical Form | Liquid |
Chemical Name or Material | Potassium iodate, Standardized Solution |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'