missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydroxide Solution, volumetric, 0.5 M KOH in methanol (0.5N), Honeywell™ Fluka™

Product Code. 15693550 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
2 L
Packaging:
Glass Bottle
Unit Size:
2 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15693550 2 L Glass Bottle 2 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15693550 Supplier Honeywell-Fluka Supplier No. 3193412L

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Volumetric, 0.5 M KOH in methanol (0.5N)

Honeywell´s high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use

 

  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations

 


 

TRUSTED_SUSTAINABILITY

Specifications

CAS 1310-58-3
Molecular Formula HKO
Linear Formula KOH
MDL Number MFCD00003553
UN Number UN2924
Density 0.83g/mL at 25°C
Synonym potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
Quantity 2 L
SMILES [OH-].[K+]
IUPAC Name potassium hydroxide
Molecular Weight (g/mol) 56.11
Flash Point 11°C (51.8°F)
PubChem CID 14797
ChEBI CHEBI:32035
Formula Weight 56.11g/mol
Packaging Glass Bottle
Chemical Name or Material Potassium Hydroxide Solution, Volumetric, 0.5M KOH in methanol (0.5N)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.