missing translation for 'onlineSavingsMsg'
Learn More

Potassium hydroxide solution, Volumetric, 0.5 M KOH in ethanol (0.5N), Honeywell Fluka™

Product Code. 15674810 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
6 x 500 mL
4 x 2.5 L
6 x 1 L
Unit Size:
1 L
2.5 L
500mL
Pack of 4
Pack of 6
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15674810 2.5 L 2.5 L
15694810 500 mL 500mL
15664810 1 L 1 L
15614820 6 x 500 mL Pack of 6
15604820 6 x 1 L Pack of 6
15684810 4 x 2.5 L Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 15674810 Supplier Honeywell-Fluka Supplier No. 351152.5L
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Volumetric, 0.5 M KOH in ethanol (0.5N)

Honeywell´s high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use

 

  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations

 


 

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-58-3
Molecular Formula HKO
Molecular Weight (g/mol) 56.11
MDL Number MFCD00003553
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
PubChem CID 14797
ChEBI CHEBI:32035
SMILES [OH-].[K+]

Specifications

Linear Formula KOH
Concentration 0.5M in ethanol
UN Number UN2924
Synonym potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
Quantity 2.5 L
IUPAC Name potassium;hydroxide
Flash Point 11°C (51.8°F)
Formula Weight 56.11g/mol
Chemical Name or Material Potassium hydroxide solution
compliance-icons compliance-icons
Product Identifier
  • Potassium hydroxide solution
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: potassium hydroxide;caustic potash, Toluene, ethanol; ethylalcohol, Hydrazinehydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.