missing translation for 'onlineSavingsMsg'
Learn More

Potassium hydroxide solution, Volumetric, 0.5 M KOH in ethanol (0.5N), Solstice

Product Code. 15604820
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
6 x 500 mL
4 x 2.5 L
6 x 1 L
Unit Size:
1 L
2.5 L
500mL
Pack of 4
Pack of 6
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15604820 6 x 1 L Pack of 6
15694810 500 mL 500mL
15664810 1 L 1 L
15674810 2.5 L 2.5 L
15614820 6 x 500 mL Pack of 6
15684810 4 x 2.5 L Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 15604820 Supplier Honeywell-Fluka Supplier No. 351156X1L
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Currently Unavailable: We have no information when this product will be available.
View alternative products
This item is not returnable. View return policy

Volumetric, 0.5 M KOH in ethanol (0.5N)

Honeywell´s high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use

 

  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations

 


 

TRUSTED_SUSTAINABILITY

Specifications

Boiling Point 78°C
CAS Min % ≥70.0000%, ≥10.0000%
CAS Max % <90.0000%, <20.0000%
CAS 7732-18-5, 1310-58-3, 108-88-3, 302-01-2
Linear Formula KOH
Concentration 0.5 M KOH in ethanol (0.5N)
Density 0.84 g/cm3
Synonym ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
Quantity 6 x 1 L
Flash Point 11°C
Grade Volumetric
Packaging Glass Bottle
Chemical Name or Material Potassium hydroxide solution, Volumetric, 0.5 M KOH in ethanol (0.5N)
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Potassium hydroxide solution
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: potassium hydroxide;caustic potash, Toluene, ethanol; ethylalcohol, Hydrazinehydrate
Product Title
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.