missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydroxide Solution 0.1M (0.1N), NIST Standard Concentrate, for Volumetric Analysis, Fisher Chemical™

Product Code. 10183711 Shop All Fisher Chemical Products
Click to view available options
Quantity:
1 Ampule
6 Ampules
Unit Size:
1 ampoule
6 ampoules
This item is not returnable. View return policy

Product Code. 10183711

Brand: Fisher Chemical J/6640C/05

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 1310-58-3
Molecular Formula HKO
Molecular Weight (g/mol) 56.11
MDL Number MFCD00003553
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
Synonym potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
PubChem CID 14797
ChEBI CHEBI:32035
SMILES [OH-].[K+]

Specifications

pH 13
Quantity 1 Ampule
IUPAC Name potassium;hydroxide
Formula Weight 56.11g/mol
Packaging Ampoule
Physical Form Liquid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.