missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydroxide in Ethanol, 0.5M, BAKER ANALYZED, J.T. Baker™

Product Code. 10138250 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
2.5 L
Packaging:
Glass
Unit Size:
2.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10138250 2.5 L Glass 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10138250 Supplier JT Baker Supplier No. 7131.2500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High quality grade volumetric solution is ready to use for laboratory and specialized industrial applications.

  • Standardized potentiometrically against an NIST Standard Reference Material, Potassium Hydrogen Phthalate
  • Label provides an actual Lot Analysis as a guarantee of purity and consistency
  • Analytical methods used for these determinations are based on standard test procedures described in technical publications
  • Improvements developed by J.T. Baker are employed to achieve a higher degree of reliability

Specifications

Color Red
Boiling Point 78°C
CAS 1310-58-3, 64-17-5
CAS Min % 0.025
Melting Point 0°C
CAS Max % 0.1, <100%
Molecular Formula HKO
Concentration 0.4975 - 0.5025mol/L KOH
MDL Number MFCD00003553
Density 0.9g/cm3 at 20°C
Synonym potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
Solubility Information Fully miscible in water
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
Quantity 2.5 L
SMILES [OH-].[K+]
IUPAC Name potassium hydroxide
Explosion Limit 3.5 - 15.0 Vol %
Molecular Weight (g/mol) 56.11
Flash Point 11°C
PubChem CID 14797
ChEBI CHEBI:32035
Formula Weight 56.11g/mol
Grade Standardized potentiometrically against a NIST Standard Reference Material, Potassium Hydrogen Phtalate
Normality 0.5N
Odor Alcohol-like
Packaging Glass
Physical Form Liquid
Chemical Name or Material Potassium Hydroxide
Show More Show Less
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.