missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium hydroxide, ACS Reagent, pellets, ≥85%, Honeywell Fluka™
57.54 GBP valid until 2024-05-31
Use promo code "21664" to get your promotional price.
Use promo code "21664" to get your promotional price.
Click to view available options
Quantity:
25 g
500 g
1 kg
2.5 kg
6 x 500 g
12 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Linear Formula | KOH |
UN Number | 1813 |
Quantity | 1 kg |
IUPAC Name | potassium;hydroxide |
Formula Weight | 56.11g/mol |
Percent Purity | ≥85% |
Grade | ACS Reagent |
Vapor Pressure | 1mmHg (719°C) |
Chemical Name or Material | Potassium Hydroxide |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'