missing translation for 'onlineSavingsMsg'
Learn More

Potassium hydroxide, Acculute Standard Volumetric Solution, Final Concentration 0.5N

Product Code. 11369309
Change view
Click to view available options
Quantity:
1 U
6 U
Unit Size:
1 unit
6 units
Product Code. Quantity unitSize
11369309 6 U 6 units
11379309 1 U 1 unit
2 options
This item is not returnable. View return policy

Product Code. 11369309

Brand: Thermo Scientific Alfa Aesar 088602.LS

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1310-58-3 | HKO | 56.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-58-3
Molecular Formula HKO
Molecular Weight (g/mol) 56.11
MDL Number MFCD00003553
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
Synonym potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
PubChem CID 14797
ChEBI CHEBI:32035
IUPAC Name potassium;hydroxide
SMILES [OH-].[K+]

Specifications

Color Colorless
UN Number UN1814
Formula Weight 56.11
Quantity 6 U
Physical Form Liquid
Chemical Name or Material Potassium hydroxide Acculute Standard Volumetric Solution, Final Concentration 0.5N
What are the instructions for dilution of 1 unit of Potassium hydroxide, Acculute Standard Volumetric Solution, Final Concentration 0.5N (Cat. No. 088602.LS, 088602.LQ)?

The following is the dilution protocol:
- Transfer entire contents of 1 unit bottle into a 1L volumetric flask.
- Rinse bottle residue, the threads of the bottle, and inner bottle cap each with DI water, and transfer to the 1L flask.
- Repeat rinse three times. Bring the 1L flask to volume with DI water and mix well.
The final solution in the 1L flask has a concentration of 0.5N Potassium hydroxide.

What is 1 unit of Potassium hydroxide, Acculute Standard Volumetric Solution, Final Concentration 0.5N (Cat. No. 088602.LS, 088602.LQ)?

1 unit of Potassium hydroxide, Acculute Standard Volumetric Solution, Final Concentration 0.5N, when diluted to 1 L, will yield a final concentration of 0.5N Potassium hydroxide solution.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.