Learn More
Potassium hydroxide, Acculute Standard Volumetric Solution, Final Concentration 0.1N, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Colorless |
| UN Number | UN1814 |
| IUPAC Name | potassium hydroxide |
| Formula Weight | 56.11 |
| Quantity | 6 U |
| Physical Form | Liquid |
| Chemical Name or Material | Potassium hydroxide Acculute Standard Volumetric Solution, Final Concentration 0.1N |
Frequently Asked Questions (FAQs)
The following is the dilution protocol:
- Transfer entire contents of 1 unit bottle into a 1L volumetric flask.
- Rinse bottle residue, the threads of the bottle, and inner bottle cap each with DI water, and transfer to the 1L flask.
- Repeat rinse three times. Bring the 1L flask to volume with DI water and mix well.
The final solution in the 1L flask has a concentration of 0.1N Potassium hydroxide.
1 unit of Potassium hydroxide, Acculute Standard Volumetric Solution, Final Concentration 0.1N, when diluted to 1 L, will yield a final concentration of 0.1N Potassium hydroxide solution.
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.