missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium hydroxide, 99.98%, (trace metal basis), pellets, Thermo Scientific Chemicals
254.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Use promo code "22914" to get your promotional price.
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
Specifications
Color | White |
Boiling Point | 1320.0°C |
Melting Point | 360.0°C |
Assay Percent Range | 99.98% |
Linear Formula | KOH |
Merck Index | 15,7761 |
Solubility Information | Solubility in water: 1120g/L (20°C). Other solubilities: 1780g/L water (100°C),soluble in ethanol (400g/L),soluble in ammonia,slightly soluble in ether |
Quantity | 500 g |
Formula Weight | 56.11 |
Percent Purity | 99.98% |
Show More |
Safety and Handling
Product Identifier
- Potassium hydroxide
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H302-Harmful if swallowed.
- H290-May be corrosive to metals.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'