missing translation for 'onlineSavingsMsg'
Learn More

Potassium hydroxide, 99.98% (metals basis), 85% min

Product Code. 11329808
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11329808 500 g 500g
11339808 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11329808 Supplier Thermo Scientific Alfa Aesar Supplier No. 044273.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1310-58-3 | HKO | 56.11 g/mol

Potassium hydroxide acts as the precursor for most of the soft and liquid soaps, and for numerous potassium-containing chemicals. It is widely used in the manufacture of detergents, fertilizer, rayon, paper, dyestuffs, various drugs, drug intermediates, fine chemicals, and petroleum products. In the field of organic chemistry, potassium hydroxide acts as a base, and is used in the synthesis of several organic compounds. Further it finds application as an electrolyte in alkaline batteries, which are based on nickel-cadmium, nickel-hydrogen and manganese dioxide-zinc.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-58-3
Molecular Formula HKO
Molecular Weight (g/mol) 56.11
MDL Number MFCD00003553
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
Synonym potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
PubChem CID 14797
ChEBI CHEBI:32035
IUPAC Name potassium;hydroxide
SMILES [OH-].[K+]

Specifications

Boiling Point 1320°C to 1324°C
Melting Point 360°C
Color White
Physical Form Pellets
Quantity 500 g
Assay Percent Range 99.98% (metals basis)
Solubility Information Freely soluble in water. Soluble in ethanol.
Formula Weight 56.11
Odor Odorless
Density 2.044
Chemical Name or Material Potassium hydroxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.