missing translation for 'onlineSavingsMsg'
Learn More

Potassium hydroxide, 1N solution in ethanol, Thermo Scientific Chemicals

Brand:  Thermo Scientific Acros 393320010

Code : 29

Additional Details : CAS Number : 64-17-5 Weight : 0.86000kg Transport : UN number : 2924 Chem class : 3 Pack group : II


 View more versions of this product

Product Code. 10153484

  • £58.10 / 1 L
Estimated Shipment: 03-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Potassium hydroxide
1N solution in ethanol
1310-58-3,64-17-5
HKO
MFCD00003553
0.8600g/mL
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
1 L
[OH-].[K+]
56.11
14797
56.11
0.86
Brown-Orange to Yellow
78.0°C
-117.0°C
KOH
0.950 to 1.050N
15, 7761
Solubility in water: soluble.
KWYUFKZDYYNOTN-UHFFFAOYSA-M
potassium hydroxide
12°C
CHEBI:32035
Plastic bottle
Liquid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Potassium hydroxide, 1N solution in ethanol

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H290-May be corrosive to metals.
  • H225-Highly flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS Documents

Documents

Certificates
Special Offers

Special Offers

For Manufacturing and Laboratory Use. Not for beverage use