missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydrogenfluoride, 99%, Honeywell Fluka™

Product Code. 15672440 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
25 g
500 g
2.5 kg
Packaging:
Drum with inliner
Poly Bottle
Unit Size:
2.5kg
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15672440 500 g Poly Bottle 500g
15662440 25 g Drum with inliner 25g
15652440 2.5 kg Poly Bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 15672440 Supplier Honeywell-Fluka Supplier No. 239283500G
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

0.99

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-29-9
MDL Number MFCD00011403

Specifications

Boiling Point 180°C
Melting Point 239°C
Quantity 500 g
Linear Formula KHF2
UN Number UN1811
Formula Weight 78.1g/mol
Percent Purity 99%
Packaging Poly Bottle
Density 2.37g/mL at 25°C
Chemical Name or Material Potassium Hydrogenfluoride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.