missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydrogen Sulfate, TRC

Product Code. 30415402
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30415402 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30415402 Supplier Toronto Research Chemicals Supplier No. TRCP6990805G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Potassium Hydrogen Sulfate
CAS 7646-93-7
Quantity 5 g
Molecular Formula HKO4S
Synonym Hydrogen Potassium Sulfate, Monopotassium Sulfate, Potassium Acid Sulfate, Potassium Bisulfate, Potassium bisulphate, Potassium Hydrogen Sulfate, Potassium Sulfate (KHSO4), Sal Enixum
InChI Formula InChI=1S/K.H2O4S/c;1-5(2,3)4/h;(H2,1,2,3,4)/q+1;/p-1
SMILES [K+].OS(=O)(=O)[O-]
IUPAC Name potassium;hydrogen sulfate
Molecular Weight (g/mol) 136.16
Formula Weight 135.9233
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.