missing translation for 'onlineSavingsMsg'
Learn More

Potassium hydrogen sulfate, 97%

Product Code. 11459633
Change view
Click to view available options
Quantity:
10,000 g
100 g
2500 g
500 g
Unit Size:
10000g
100g
2500g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11459633 10,000 g 10000g
11434020 100 g 100g
11439633 500 g 500g
11449633 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11459633 Supplier Thermo Scientific Alfa Aesar Supplier No. A12428.0C
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 7646-93-7 | HKO4S | 136.16 g/mol

Potassium hydrogen sulfate is used in the conversion of tartrates to bitartrates in wine. It serves as a disintegrating agent in analytical chemistry and as an intermediate in the preparation of potassium persulfate by electrolysis. It is also employed in the manufacturing of fertilizers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium hydrogen sulfate is used in the conversion of tartrates to bitartrates in wine. It serves as a disintegrating agent in analytical chemistry and as an intermediate in the preparation of potassium persulfate by electrolysis. It is also employed in the manufacturing of fertilizers.

Solubility
Soluble in water, acetone and ethanol.

Notes
Hygroscopic. Incompatible with strong bases, strong oxidizing agents and alcohols.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7646-93-7
Molecular Formula HKO4S
Molecular Weight (g/mol) 136.16
MDL Number MFCD00011404
InChI Key CHKVPAROMQMJNQ-UHFFFAOYSA-M
Synonym potassium bisulfate, potassium hydrogen sulfate, potassium bisulphate, sulfuric acid, monopotassium salt, potassium acid sulfate, sal enixum, potassium hydrogensulphate, monopotassium sulfate, acid potassium sulfate, caswell no. 682c
PubChem CID 516920
IUPAC Name potassium;hydrogen sulfate
SMILES [K+].OS([O-])(=O)=O

Specifications

Melting Point 195°C to 197°C (decomposition)
Quantity 10,000 g
Linear Formula KHSO4
UN Number UN2509
Merck Index 14,7613
Solubility Information Soluble in water,acetone and ethanol.
Formula Weight 136.17
Percent Purity 97%
Odor Odorless
Sensitivity Hygroscopic
Density 2.24
Chemical Name or Material Potassium hydrogen sulfate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.