missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydrogen Phthalate, Certified AR for Analysis, Fisher Chemical™

Potassium Hydrogen Phthalate, Certified AR for Analysis, Fisher Chemical, Quantity: 500g, CAS: 877-24-7, White, Melting Point: 295 deg.C, Molecular Formula: C8H5KO4, Molecular Weight: 204.222, Packaging: Plastic powder jar, pH: 3.8 to 4.0

£46.00 - £224.00

Chemical Identifiers

CAS 877-24-7
Molecular Formula C8H5KO4
Molecular Weight (g/mol) 204.222
MDL Number 13070
InChI Key IWZKICVEHNUQTL-UHFFFAOYSA-M
Synonym potassium hydrogen phthalate, potassium biphthalate, monopotassium phthalate, potassium acid phthalate, hydrogen potassium phthalate, phthalic acid monopotassium salt, 1,2-benzenedicarboxylic acid, monopotassium salt, phthalic acid, monopotassium salt, phthalic acid potassium salt, potassium phthalate monobasic
PubChem CID 23676735
IUPAC Name potassium;2-carboxybenzoate
SMILES C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+]
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10254300
View Documents Promotion Details
Fisher Chemical
P/5320/48
100 g
£46.00
100g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
10040340
View Documents Promotion Details
Fisher Chemical
P/5320/50
250 g
£83.70
250g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
10437460
View Documents Promotion Details
Fisher Chemical
P/5320/53
500 g
£141.00
500g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
10151960
View Documents Promotion Details
Fisher Chemical
P/5320/60
1 kg
£224.00
1kg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Potassium Hydrogen Phthalate, Certified AR for Analysis, Fisher Chemical, Quantity: 500g, CAS: 877-24-7, White, Melting Point: 295 deg.C, Molecular Formula: C8H5KO4, Molecular Weight: 204.222, Packaging: Plastic powder jar, pH: 3.8 to 4.0

Specifications

Chemical Identifiers

877-24-7
204.222
IWZKICVEHNUQTL-UHFFFAOYSA-M
23676735
C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+]
C8H5KO4
13070
potassium hydrogen phthalate, potassium biphthalate, monopotassium phthalate, potassium acid phthalate, hydrogen potassium phthalate, phthalic acid monopotassium salt, 1,2-benzenedicarboxylic acid, monopotassium salt, phthalic acid, monopotassium salt, phthalic acid potassium salt, potassium phthalate monobasic
potassium;2-carboxybenzoate

Specifications

295°C
White
Solid
13070
IWZKICVEHNUQTL-UHFFFAOYSA-M
potassium;2-carboxybenzoate
23676735
Plastic powder jar
877-24-7
3.8 to 4.0
C8H5KO4
potassium hydrogen phthalate, potassium biphthalate, monopotassium phthalate, potassium acid phthalate, hydrogen potassium phthalate, phthalic acid monopotassium salt, 1,2-benzenedicarboxylic acid, monopotassium salt, phthalic acid, monopotassium salt, phthalic acid potassium salt, potassium phthalate monobasic
C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+]
204.222
204.22g/mol
Videos
SDS
Documents

Documents

Product Certifications