missing translation for 'onlineSavingsMsg'
Learn More

Potassium hydrogen phosphate, dried, 98+%

Product Code. 11483714
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11483714 1000 g 1000g
11493714 250 g 250g
11403724 5000 g 5000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11483714 Supplier Thermo Scientific Alfa Aesar Supplier No. A11321.0B
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7758-11-4 | HK2O4P | 174.17 g/mol

Potassium hydrogen phosphate is used as a fertilizer, food additive, buffering agent, in molecular biology, biochemistry and chromatography. It is useful to increase the partition coefficients of amino acids in aqueous two-phase systems. It plays an important role for increase in rice grain yield by spraying chemicals including ferric chloride, dipotassium hydrogen phosphate and salicylic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium hydrogen phosphate is used as a fertilizer, food additive, buffering agent, in molecular biology, biochemistry and chromatography. It is useful to increase the partition coefficients of amino acids in aqueous two-phase systems. It plays an important role for increase in rice grain yield by spraying chemicals including ferric chloride, dipotassium hydrogen phosphate and salicylic acid.

Solubility
Soluble in water. Slightly soluble in alcohol.

Notes
Hygroscopic chemical and incompatible with strong oxidizing agents and acids. Keep the material away from heat, sparks and flame and sources of ignition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 11-4-7758
Molecular Formula HK2O4P
Molecular Weight (g/mol) 174.17
MDL Number MFCD00011383
InChI Key ZPWVASYFFYYZEW-UHFFFAOYSA-L
Synonym dipotassium hydrogen phosphate, dipotassium phosphate, dipotassium hydrogenphosphate, dibasic potassium phosphate, potassium hydrogen phosphate, potassium phosphate, dibasic, potassium dibasic phosphate, potassium phosphate dibasic, phosphoric acid, dipotassium salt, dipotassium monophosphate
PubChem CID 24450
ChEBI CHEBI:32031
IUPAC Name dipotassium;hydrogen phosphate
SMILES OP(=O)([O-])[O-].[K+].[K+]

Specifications

Quantity 1000 g
Merck Index 14,7658
Solubility Information Soluble in water. Slightly soluble in alcohol.
Formula Weight 174.18
Percent Purity ≥98%
Sensitivity Hygroscopic
Chemical Name or Material Potassium hydrogen phosphate, Dried

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.