missing translation for 'onlineSavingsMsg'
Learn More

Potassium hydrogen iodate, primary standard, ACS, 99.95-100.05%

Product Code. 11344987
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11344987 25 g 25g
11334987 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11344987 Supplier Thermo Scientific Alfa Aesar Supplier No. 041986.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Maximum level of impurities: Insoluble matter 0.01%, SO4 0.005%, Fe 0.001%, Cl 0.02%, Heavy Metals (as Pb) 0.001% | CAS: 13455-24-8 | HI2KO6 | 389.909 g/mol

Potassium hydrogen iodate acts as an oxidizing agent. Further, it serves as an intermediate for the synthesis of potassium iodate and also used as an analysis reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13455-24-8
Molecular Formula HI2KO6
Molecular Weight (g/mol) 389.909
MDL Number MFCD00011400
InChI Key ACAYDTMSDROWHW-UHFFFAOYSA-M
PubChem CID 23700942
IUPAC Name potassium;iodic acid;iodate
SMILES OI(=O)=O.[O-]I(=O)=O.[K+]

Specifications

Assay Percent Range 99.95-100.05%
Physical Form Powder
Color White
Solubility Soluble in water
Quantity 25 g
Formula Weight 389.91
Grade ACS
Chemical Name or Material Potassium hydrogen iodate, Primary Standard

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.