missing translation for 'onlineSavingsMsg'
Learn More

Potassium hydrogen fluoride, 98%, Thermo Scientific Chemicals

Product Code. 11419963
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11419963 500 g 500g
11444020 100 g 100g
11429963 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11419963 Supplier Thermo Scientific Alfa Aesar Supplier No. A13031.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Potassium hydrogen fluoride is used as an abrasive, as a pretreatment of metals before they are further processed, as a wood preservative and as a chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium hydrogen fluoride is used as an abrasive, as a pretreatment of metals before they are further processed, as a wood preservative and as a chemical intermediate.

Solubility
Soluble in water (392 g/L). Also soluble in ethanol.

Notes
It is hygroscopic. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 7789-29-9
Molecular Formula KHF2
MDL Number MFCD00011403
Synonym Potassium bifluoride

Specifikationer

Melting Point 239°C (decomposition)
pH 1
Quantity 500 g
UN Number UN1811
Merck Index 14,7610
Solubility Information Soluble in water (392g/L). Also soluble in ethanol.
Formula Weight 78.1
Percent Purity 98%
Odor Odorless
Sensitivity Hygroscopic
Density 2.37
Chemical Name or Material Potassium hydrogen fluoride
Visa mer Visa mindre
compliance-icons compliance-icons
Produktbeteckning
  • Potassium hydrogen fluoride
Signalord
  • Danger
Riskkategorier
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Faroangivelser
  • H301-Toxic if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Skyddsangivelse
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Kompletterande information
  • MIXTURE LIST-Contain: Potassium fluoride

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.