missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium hydrogen carbonate, extra pure
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 kg
5 kg
25 kg
Packaging:
Fibre drum
Plastic bottle
Unit Size:
1kg
25kg
5kg
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 292.0°C |
| Quantity | 5 kg |
| Linear Formula | KHCO3 |
| Merck Index | 15,773 |
| Solubility Information | Solubility in water: 333g/L (20°C). Other solubilities: 600g/L water (60°C), practocally insoluble in alcohol, soluble in potassium carbonate solution |
| IUPAC Name | potassium;hydrogen carbonate |
| Formula Weight | 100.12 |
| Grade | Extra Pure |
| Packaging | Plastic bottle |
| Chemical Name or Material | Potassium hydrogen carbonate |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction