missing translation for 'onlineSavingsMsg'
Learn More

Potassium hydrogen carbonate, 99%

Product Code. 11439010
Change view
Click to view available options
Quantity:
10,000 g
2500 g
500 g
Unit Size:
10000g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11439010 2500 g 2500g
11449010 500 g 500g
11429010 10,000 g 10000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11439010 Supplier Thermo Scientific Alfa Aesar Supplier No. A12429.0E
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 298-14-6 | CHKO3 | 100.11 g/mol

Potassium hydrogen carbonate is used in foods to regulate pH, as a fire suppression agent (BC dry chemical) and as a buffering agent in medications. It acts as an additive in winemaking and is an active component in club soda. Furthermore, it finds use in crops for the neutralization of acidic soils. It is also used as a source of carbon dioxide for leavening in baking. It is utilized to inhibit the growth of aspergillus parasiticus in czapek's agar. It is employed as a catalyst in synthetic fiber polymerization and olefin dehydrogenation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium hydrogen carbonate is used in foods to regulate pH, as a fire suppression agent (BC dry chemical) and as a buffering agent in medications. It acts as an additive in winemaking and is an active component in club soda. Furthermore, it finds use in crops for the neutralization of acidic soils. It is also used as a source of carbon dioxide for leavening in baking. It is utilized to inhibit the growth of aspergillus parasiticus in czapek′s agar. It is employed as a catalyst in synthetic fiber polymerization and olefin dehydrogenation.

Solubility
Soluble in water. Insoluble in alcohol.

Notes
Incompatible with strong oxidizing agents and strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 298-14-6
Molecular Formula CHKO3
Molecular Weight (g/mol) 100.11
MDL Number MFCD00011402
InChI Key TYJJADVDDVDEDZ-UHFFFAOYSA-M
Synonym potassium bicarbonate, potassium hydrogen carbonate, potassium hydrogencarbonate, potassium acid carbonate, monopotassium carbonate, carbonic acid, monopotassium salt, unii-hm5z15lebn, potassiumbicarbonate, ccris 3510, khco3
PubChem CID 516893
ChEBI CHEBI:81862
IUPAC Name potassium;hydrogen carbonate
SMILES [K+].OC([O-])=O

Specifications

Quantity 2500 g
Merck Index 14,7609
Solubility Information Soluble in water. Insoluble in alcohol.
Formula Weight 100.12
Percent Purity 99%
Odor Odorless
Sensitivity Hygroscopic
Density 2.17
Chemical Name or Material Potassium hydrogen carbonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.