missing translation for 'onlineSavingsMsg'
Learn More

Potassium hexahydroxyplatinate(IV), Premion™, 99.95% (metals basis), Pt 51.5% min

Product Code. 11359417
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11359417 5 g 5g
11369417 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11359417 Supplier Thermo Scientific Alfa Aesar Supplier No. 041014.06
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 12285-90-4 | H12K2O6Pt+2 | 381.371 g/mol

Potassium hexahydroxyplatinate(IV) is an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12285-90-4
Molecular Formula H12K2O6Pt+2
Molecular Weight (g/mol) 381.371
MDL Number MFCD00064671
InChI Key LTROBPQZKJGHPB-UHFFFAOYSA-N
Synonym potassium hexahydroxoplatinate, dipotassium platinate, potassium hexahydroxyplatinate iv, platinum potassium hydroxide, h6k2o6pt, potassium hexahydroxoplatinate iv, potassium hexahydroxyplatinum iv, potassium hexahydroxyplatinate, dipotassium hexahydroxyplatinumdiuide, premion metalsbasis ,ptmin
PubChem CID 16717651
IUPAC Name dipotassium;platinum;hexahydrate
SMILES O.O.O.O.O.O.[K+].[K+].[Pt]

Specifications

Physical Form Powder
Quantity 5 g
Solubility Information Soluble in water.
Formula Weight 375.34
Percent Purity 99.95%
Grade Premion
Concentration or Composition (by Analyte or Components) Pt 51.5% min.
Assay (metals basis)
Chemical Name or Material Potassium hexahydroxyplatinate(IV)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.