missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hexahydroxyantimonate (KSb(OH)6), TRC

Product Code. 30421276
Change view
Click to view available options
Quantity:
250 mg
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30421276 5 g
30416108 250 mg
30413650 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30421276 Supplier Toronto Research Chemicals Supplier No. TRCP6987285G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12208-13-8
Molecular Formula H6KO6Sb
Molecular Weight (g/mol) 262.9
Synonym Potassium Antimonate(V) (K[Sb(OH)6]), Antimony Potassium Hydroxide (SbK(OH)6), Potassium Antimony Hydroxide (KSb(OH)6), Potassium Hexahydroxoantimonate(1-), Antimonic Acid Monopotassium Salt
IUPAC Name potassium;hexahydroxyantimony(1-)
SMILES [K+].O[Sb-](O)(O)(O)(O)O

Specifications

Quantity 5 g
InChI Formula InChI=1S/K.6H2O.Sb/h;6*1H2;/q+1;;;;;;;+5/p-6
Formula Weight 261.884
Chemical Name or Material Potassium Hexahydroxyantimonate (KSb(OH)6)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.