missing translation for 'onlineSavingsMsg'
Learn More

Potassium hexafluorozirconate, 99%, Thermo Scientific Chemicals

Product Code. 11334939
Change view
Click to view available options
Quantity:
500 g
2 kg
Unit Size:
2kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11334939 2 kg 2kg
11344939 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11334939 Supplier Thermo Scientific Alfa Aesar Supplier No. 086102.A3
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For manufacture of metallic zirconium

Employed in the manufacture of metallic zirconium. Used in the production of aluminum magnesium alloy, ceramics and glass.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16923-95-8
Molecular Formula F6K2Zr
Molecular Weight (g/mol) 283.411
MDL Number MFCD00042547
InChI Key BJZIJOLEWHWTJO-UHFFFAOYSA-H
Synonym potassium hexafluorozirconate, zirconium potassium fluoride, potassium fluorozirconate, f6zr.2k, dipotassium hexafluorozirconate, potassium hexafluorozirconate iv, dipotassium hexafluorozirconiumdiuide, dipotassium hexafluorozirconate iv
PubChem CID 56828182
IUPAC Name dipotassium;hexafluorozirconium(2-)
SMILES F[Zr-2](F)(F)(F)(F)F.[K+].[K+]

Specifications

Quantity 2 kg
UN Number UN3288
Merck Index 14,7639
Solubility Information Slightly soluble in cold water, soluble in hot water
Formula Weight 283.41
Percent Purity 99%
Odor Odorless
Density 3.48 g/mL
Chemical Name or Material Potassium hexafluorozirconate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.