missing translation for 'onlineSavingsMsg'
Learn More

Potassium hexafluorosilicate, 99.999% (metals basis)

Product Code. 11475200
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11475200 250 g 250g
11455200 10 g 10g
11465200 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11475200 Supplier Thermo Scientific Alfa Aesar Supplier No. 044495.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 16871-90-2 | F6K2Si | 220.27 g/mol

Potassium hexafluorosilicate is used in the production of porceleins. It is also used for the preservation of timber. It is involved in the manufacturing of ceramics, aluminum and magnesium smelting. In addition, it is used as an intermediate in the manufacturing of optical glass.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16871-90-2
Molecular Formula F6K2Si
Molecular Weight (g/mol) 220.27
MDL Number MFCD00040666
InChI Key UUHPPUWEDCVPCO-UHFFFAOYSA-N
Synonym potassium hexafluorosilicate, potassium fluosilicate, potassium silicofluoride, dipotassium hexafluorosilicate, silicate 2-, hexafluoro-, dipotassium, unii-2610w49xyd, dipotassium hexafluorosilicon 2-, dipotassium hexafluorosilicate 2-, sif6.2k, potassium hexafluorosilicate iv
PubChem CID 61851
IUPAC Name dipotassium;hexafluorosilicon(2-)
SMILES [K+].[K+].F[Si--](F)(F)(F)(F)F

Specifications

Melting Point (decomposition)
Color White
pH 3.4
Physical Form Crystalline
Quantity 250 g
Linear Formula K2SiF6
UN Number UN2655
Merck Index 14,7638
Solubility Information Slightly soluble in water.
Formula Weight 220.28
Percent Purity 99.999%
Sensitivity Moisture sensitive
Density 2.27 g/mL
Assay (metals basis)
Chemical Name or Material Potassium hexafluorosilicate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.