missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hexafluorophosphate, TRC

Product Code. 30410630
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30410630 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30410630 Supplier Toronto Research Chemicals Supplier No. TRCP698958250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Potassium Hexafluorophosphate
CAS 17084-13-8
Quantity 250 mg
Molecular Formula F6 P . K
Synonym Monopotassium Hexafluorophosphate, NSC 404039, Potassium Fluophosphate, Potassium Fluorophosphate, Potassium Hexafluorophosphate (KPF6), Potassium Hexafluorophosphate(1-)
InChI Formula InChI=1S/F6P.K/c1-7(2,3,4,5)6;/q-1;+1
SMILES [K+].F[P-](F)(F)(F)(F)F
IUPAC Name potassium;hexafluorophosphate
Molecular Weight (g/mol) 184.062
Formula Weight 183.928
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.