missing translation for 'onlineSavingsMsg'
Learn More

Potassium hexafluorophosphate, 99%, extra pure, Thermo Scientific™

Click to view available options
Quantity:
50g
250g
1kg
2.5kg
Packaging:
Plastic bottle
This item is not returnable. View return policy
17084-13-8
F6KP
184.07
MFCD00011412
YZDGRYDIGCWVND-UHFFFAOYSA-N
potassium hexafluorophosphate, potassium fluophosphate, potassium hexafluorophosphate v, unii-88w436pdi9, monopotassium hexafluorophosphate, potassium hexfluorophosphate, potassium hexafluorophosphate min, phosphate 1-, hexafluoro-, potassium, potassium fluorophosphate, kpf6
23688904
potassium;hexafluorophosphate
F[P-](F)(F)(F)(F)F.[K+]
This item is not returnable. View return policy
17084-13-8
F6KP
184.07
MFCD00011412
YZDGRYDIGCWVND-UHFFFAOYSA-N
potassium hexafluorophosphate, potassium fluophosphate, potassium hexafluorophosphate v, unii-88w436pdi9, monopotassium hexafluorophosphate, potassium hexfluorophosphate, potassium hexafluorophosphate min, phosphate 1-, hexafluoro-, potassium, potassium fluorophosphate, kpf6
23688904
potassium;hexafluorophosphate
F[P-](F)(F)(F)(F)F.[K+]

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Color White
Melting Point 575.0°C
Quantity 1kg
Assay Percent Range 98.5% min
Linear Formula KPF6
Solubility Information Solubility in water: 93g/L (25°C)
Formula Weight 184.07
Physical Form Crystalline Powder
Percent Purity 99%
Grade Extra Pure
Loss on Drying 0.1% max. (100°C)
Packaging Plastic bottle
Solubility Solubility in water: 93g/L (25°C).
Additional Information free fluoride (F): < 0.5%
Chemical Name or Material Potassium hexafluorophosphate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Potassium hexafluorophosphate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only