missing translation for 'onlineSavingsMsg'
Learn More

Potassium hexafluoroarsenate(V), 99% (metals basis), Thermo Scientific™

Product Code. p-4824891
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Product Code. 11397217

Brand: Thermo Scientific Alfa Aesar 011532.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17029-22-0
Molecular Formula AsF6K
Molecular Weight (g/mol) 228.01
MDL Number MFCD00042546
InChI Key DITNVAZRXJOPSJ-UHFFFAOYSA-N
Synonym potassium hexafluoroarsenate, hexaflurate, nopalmitate, nopalmate, caswell no. 692b, potassium hexafluoroarsenate v, unii-t7tyz7585f, potassium hexafluoroarsenate 1-, arsenate 1-, hexafluoro-, potassium, epa pesticide chemical code 599700
PubChem CID 159810
ChEBI CHEBI:82182
SMILES [K+].F[As-](F)(F)(F)(F)F

Specifications

Melting Point 400°C
Color White
Physical Form Powder
Quantity 5 g
Assay Percent Range 99% (metals basis)
IUPAC Name potassium hexafluoro-λ⁵-arsanuide
Formula Weight 228.01
Odor Acrid
Appearance White powder
Chemical Name or Material Potassium hexafluoroarsenate(V)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.