missing translation for 'onlineSavingsMsg'
Learn More

Potassium hexafluoroarsenate(V), 99% (metals basis), Thermo Scientific™

Catalog Number 11397217
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
17029-22-0
AsF6K
228.01
MFCD00042546
DITNVAZRXJOPSJ-UHFFFAOYSA-N
potassium hexafluoroarsenate, hexaflurate, nopalmitate, nopalmate, caswell no. 692b, potassium hexafluoroarsenate v, unii-t7tyz7585f, potassium hexafluoroarsenate 1-, arsenate 1-, hexafluoro-, potassium, epa pesticide chemical code 599700
159810
CHEBI:82182
[K+].F[As-](F)(F)(F)(F)F
This item is not returnable. View return policy
17029-22-0
AsF6K
228.01
MFCD00042546
DITNVAZRXJOPSJ-UHFFFAOYSA-N
potassium hexafluoroarsenate, hexaflurate, nopalmitate, nopalmate, caswell no. 692b, potassium hexafluoroarsenate v, unii-t7tyz7585f, potassium hexafluoroarsenate 1-, arsenate 1-, hexafluoro-, potassium, epa pesticide chemical code 599700
159810
CHEBI:82182
[K+].F[As-](F)(F)(F)(F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 400°C
Color White
Physical Form Powder
Quantity 5 g
Assay Percent Range 99% (metals basis)
IUPAC Name potassium hexafluoro-λ⁵-arsanuide
Formula Weight 228.01
Odor Acrid
Appearance White powder
Chemical Name or Material Potassium hexafluoroarsenate(V)
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'