missing translation for 'onlineSavingsMsg'
Learn More

Potassium hexafluoroarsenate(V), 99% (metals basis), Thermo Scientific™

Product Code. 11307227
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17029-22-0
Molecular Formula AsF6K
Molecular Weight (g/mol) 228.01
MDL Number MFCD00042546
InChI Key DITNVAZRXJOPSJ-UHFFFAOYSA-N
Synonym potassium hexafluoroarsenate, hexaflurate, nopalmitate, nopalmate, caswell no. 692b, potassium hexafluoroarsenate v, unii-t7tyz7585f, potassium hexafluoroarsenate 1-, arsenate 1-, hexafluoro-, potassium, epa pesticide chemical code 599700
PubChem CID 159810
ChEBI CHEBI:82182
SMILES [K+].F[As-](F)(F)(F)(F)F

Specifications

Melting Point 400°C
Color White
Physical Form Powder
Quantity 25 g
Assay Percent Range 99% (metals basis)
IUPAC Name potassium;hexafluoroarsenic(1-)
Formula Weight 228.01
Odor Acrid
Appearance White powder
Chemical Name or Material Potassium hexafluoroarsenate(V)
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'