missing translation for 'onlineSavingsMsg'
Learn More

Potassium hexacyanoruthenate(II) hydrate, Ru 23.0% min

Product Code. 11331958
Change view
Click to view available options
Quantity:
0.2 g
2 g
10 g
Unit Size:
0.2g
10g
2g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11331958 0.2 g 0.2g
11321958 2 g 2g
11341958 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11331958 Supplier Thermo Scientific Alfa Aesar Supplier No. 011876.SS
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 339268-21-2 | C6K4N6Ru | 413.57 g/mol

Potassium hexacyanoruthenate(II) hydrate used in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 339268-21-2
Molecular Formula C6K4N6Ru
Molecular Weight (g/mol) 413.57
MDL Number MFCD00168064
InChI Key FULSJSKUKUZHLZ-UHFFFAOYSA-N
Synonym potassium hexacyanoruthenate ii hydrate, tetrapotassium hexacyanorutheniumtetrauide hydrate
PubChem CID 71311369
IUPAC Name tetrapotassium;ruthenium(2+);hexacyanide;hydrate
SMILES [K+].[K+].[K+].[K+].N#C[Ru++](C#N)(C#N)(C#N)(C#N)C#N

Specifications

Color White
Physical Form Crystalline
Quantity 0.2 g
Linear Formula K4Ru(CN)6·xH2O
UN Number UN1588
Solubility Information Soluble in water.
Formula Weight 413.57 (Anhydrous)
Concentration or Composition (by Analyte or Components) Ru 23.0% min.
Chemical Name or Material Potassium hexacyanoruthenate(II) hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.