missing translation for 'onlineSavingsMsg'
Learn More

Potassium hexacyanoferrate(III), ACS, 99.0% min

Product Code. p-4782076
Click to view available options
Quantity:
500 g
2 kg
Unit Size:
2kg
500g
This item is not returnable. View return policy

Product Code. 11307688

Brand: Thermo Scientific Alfa Aesar 033357.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13746-66-2 | C6FeK3N6 | 329.25 g/mol

Potassium hexacyanoferrate(III) is used in reading palimpsests and old manuscripts, in blueprint drawing and in photography (Cyanotype process). It finds application to harden iron and steel, in electroplating, dyeing wool and as a laboratory reagent. It is a mild oxidizing agent used in organic synthesis. It is associated with potassium hydroxide solution used to formulate Murakami’s etchant, which is useful for metallographers to provide contrast between binder and carbide phases in cemented carbides. Further, it is used in many amperometric biosensors as an electron transfer agent replacing an enzyme's natural electron transfer agent such as oxygen with the enzyme glucose oxidase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13746-66-2
Molecular Formula C6FeK3N6
Molecular Weight (g/mol) 329.25
MDL Number MFCD00011392
InChI Key QMTKJUMXUDIUAQ-UHFFFAOYSA-N
Synonym potassium ferricyanide, red prussiate, potassium ferricyanate, tripotassium hexacyanoferrate, potassium cyanoferrate, iron potassium cyanide, tripotassium ferricyanide, tripotassium iron hexacyanide, tripotassium ferric hexacyanide, ccris 5559
PubChem CID 26250
IUPAC Name tripotassium;iron(3+);hexacyanide
SMILES [K+].[K+].[K+].N#C[Fe+3](C#N)(C#N)(C#N)(C#N)C#N

Specifications

Melting Point (decomposition)
Color Orange
Physical Form Crystalline
Quantity 500 g
Assay Percent Range 99.0% min.
Solubility Information Soluble in water and acid. Slightly soluble in alcohol.
Formula Weight 329.26
Grade ACS
Sensitivity Light sensitive
Appearance Orange crystalline
Density 1.89 g/cm3 at 20°C
Chemical Name or Material Potassium hexacyanoferrate(III)
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.