missing translation for 'onlineSavingsMsg'
Learn More

Potassium hexacyanocobaltate(III), 90+%

Product Code. 11332608
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11332608 250 g 250g
11342608 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11332608 Supplier Thermo Scientific Alfa Aesar Supplier No. 023126.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13963-58-1

It is applied in formation of a xerogel via a cyanogel, the gel has potential use in solid-state gas sensors. It is also used as reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13963-58-1
MDL Number MFCD00011391

Specifications

Melting Point (decomposition)
Color Brown to Yellow
Physical Form Crystalline
Quantity 250 g
Linear Formula K3[Co(CN)6]
Merck Index 14,7637
Solubility Information Soluble in water and acetic acid solutions; Insoluble in alcohol
Formula Weight 332.33
Percent Purity ≥90%
Odor Odorless
Sensitivity Light sensitive
Density 1.906 g/mL
Chemical Name or Material Potassium hexacyanocobaltate(III)
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.