missing translation for 'onlineSavingsMsg'
Learn More

Potassium hexachlororuthenate(IV), Premion™, 99.95% (metal basis), Ru 25.3% min

Product Code. 11383988
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11383988 1 g 1g
11373988 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11383988 Supplier Thermo Scientific Alfa Aesar Supplier No. 040399.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 23013-82-3 | K2RuCl6

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23013-82-3
Molecular Formula K2RuCl6
MDL Number MFCD00050159

Specifications

Color Red to Brown
Physical Form Crystalline Powder
Quantity 1 g
Assay Percent Range 99.95% (metals basis)
Formula Weight 391.99
Appearance Red-browm crystalline powder
Chemical Name or Material Potassium hexachlororuthenate(IV)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.