missing translation for 'onlineSavingsMsg'
Learn More

Potassium hexachlororuthenate(III), Premion™, 99.99% (metals basis), Ru 23% min

Product Code. 11495300
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11495300 1 g 1g
11405310 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11495300 Supplier Thermo Scientific Alfa Aesar Supplier No. 044547.03
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 25443-63-4 | K3RuCl6

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25443-63-4
Molecular Formula K3RuCl6
MDL Number MFCD00011390
Synonym ruthenium 6+ hexachloride, tripotassium hexachlororuthenate, ruthenium hexachloride, ruthenium chloride rucl6 , oc-6-11, ruthenate 3-, tripotassium, oc-6-11

Specifications

Physical Form Crystalline
Quantity 1 g
Formula Weight 431.09
Percent Purity 99.99%
Grade Premion
Concentration or Composition (by Analyte or Components) Ru 23% min
Assay (metals basis)
Chemical Name or Material Potassium hexachlororuthenate (III)
compliance-icons
Product Identifier
  • Potassium hexachlororuthanate(III)
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Tripotassium hexachlororuthenate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.