missing translation for 'onlineSavingsMsg'
Learn More

Potassium hexachlororhodate(III), Premion™, 99.99% (metals basis), Rh 23.3% min

Product Code. 11349417
Change view
Click to view available options
Quantity:
0.25 g
1 g
5 g
Unit Size:
0.25g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11349417 0.25 g 0.25g
11329417 1 g 1g
11339417 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11349417 Supplier Thermo Scientific Alfa Aesar Supplier No. 041013.01
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

CAS: 13845-07-3 | Cl6K3Rh | 432.89 g/mol

Potassium hexachlororhodate(III) is used as pharmaceutical intermediates. It is also used as chemical catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13845-07-3
Molecular Formula Cl6K3Rh
Molecular Weight (g/mol) 432.89
MDL Number MFCD00192414
InChI Key MJKHGKZFPXUBSV-UHFFFAOYSA-H
Synonym potassium hexachlororhodate iii, tripotassium;hexachlororhodium 3-, tripotassium hexachlororhodate 3-, potassium hexachloro-rhodate-iii ,hydrate
PubChem CID 56923611
SMILES [K+].[K+].[K+].Cl[Rh+7](Cl)(Cl)(Cl)(Cl)Cl

Specifications

Physical Form Powder
Quantity 0.25 g
Solubility Information Slightly soluble in cold water,insoluble in alcohol.
IUPAC Name rhodium(3+) tripotassium hexachloride
Formula Weight 432.93
Percent Purity 99.99%
Grade Premion
Concentration or Composition (by Analyte or Components) Rh 23.3% min.
Sensitivity Hygroscopic
Assay (metals basis)
Chemical Name or Material Potassium hexachlororhodate(III)
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.