missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hexachloroplatinate(Iv), Extra Pure, SLR, Fisher Chemical

Product Code. 10785911 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Vial
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10785911 1 g Vial 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10785911 Supplier Fisher Chemical Supplier No. P/4320/43
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Melting Point 250°C
CAS 16921-30-5
Color Orange to Yellow
Physical Form Solid
Quantity 1 g
Molecular Formula Cl6K2Pt
MDL Number MFCD00011389
Synonym potassium hexachloroplatinate iv, potassium platinic chloride, potassium hexachloroplatinate, dipotassium platinum hexachloride, platinic potassium chloride, dipotassium hexachloroplatinate, potassium platinum chloride, dipotassium hexachloroplatinate 2-, ccris 7824, platinum potassium chloride
InChI Key DPAIVKJGTXERIM-UHFFFAOYSA-H
SMILES [K+].[K+].Cl[Pt--](Cl)(Cl)(Cl)(Cl)Cl
IUPAC Name dipotassium hexachloroplatinumdiuide
Molecular Weight (g/mol) 485.98
PubChem CID 61856
Formula Weight 486.01g/mol
Packaging Vial
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.