missing translation for 'onlineSavingsMsg'
Learn More

Potassium hexachloroplatinate(IV), ca. 40% Pt

Product Code. 10216350 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10216350 5 g Glass bottle 5g
10348000 1 g Glass bottle 1g
11944341 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10216350 Supplier Thermo Scientific Acros Supplier No. 195350050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

CAS: 16921-30-5 | Cl6K2Pt | 485.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16921-30-5
Molecular Formula Cl6K2Pt
Molecular Weight (g/mol) 485.98
MDL Number MFCD00011389
InChI Key DPAIVKJGTXERIM-UHFFFAOYSA-H
Synonym potassium hexachloroplatinate iv, potassium platinic chloride, potassium hexachloroplatinate, dipotassium platinum hexachloride, platinic potassium chloride, dipotassium hexachloroplatinate, potassium platinum chloride, dipotassium hexachloroplatinate 2-, ccris 7824, platinum potassium chloride
PubChem CID 61856
SMILES [K+].[K+].Cl[Pt--](Cl)(Cl)(Cl)(Cl)Cl

Specifications

Melting Point 250.0°C
Color Black
Physical Form Powder
Quantity 5 g
Linear Formula K2PtCl6
Merck Index 15,7757
Solubility Information Solubility in water: 50g/L (95°C). Other solubilities: insoluble in alcohol
IUPAC Name dipotassium;hexachloroplatinum(2-)
Formula Weight 486.01
Percent Purity ∼40% (Pt)
Packaging Glass bottle
Chemical Name or Material Potassium hexachloroplatinate(IV)
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Potassium hexachloroplatinate(IV)
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 1
Hazard Statement
  • H290-May be corrosive to metals.
  • H301-Toxic if swallowed.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H372-Causes damage to organs.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.