missing translation for 'onlineSavingsMsg'
Learn More

Potassium hexachloropalladate(IV), Pd 26.3% min

Product Code. 11381968
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11381968 1 g 1g
11391968 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11381968 Supplier Thermo Scientific Alfa Aesar Supplier No. 011884.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 16919-73-6 | Cl6K2Pd | 397.317 g/mol

Potassium hexachloropalladate(IV) is used as organic reagents and pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16919-73-6
Molecular Formula Cl6K2Pd
Molecular Weight (g/mol) 397.317
MDL Number MFCD00043081
InChI Key OEDRXJBJMMJUSV-UHFFFAOYSA-H
Synonym potassium hexachloropalladate iv, dipotassium hexachloropalladate, dipotassium hexachloropalladate 2-, ccris 7827, potassiuim hexachloropalladate iv, palladate 2-, hexachloro-, dipotassium, potassium palladium iv chloride, palladate 2-, hexachloro-, dipotassium, oc-6-11, potassium chloropalladate iv 6ci, dipotassiumhexachloropalladate
PubChem CID 61852
IUPAC Name dipotassium;hexachloropalladium(2-)
SMILES Cl[Pd-2](Cl)(Cl)(Cl)(Cl)Cl.[K+].[K+]

Specifications

Melting Point (decomposition)
Color Red
Physical Form Crystalline
Quantity 1 g
UN Number UN3077
Solubility Information Soluble in water.
Formula Weight 397.32
Concentration or Composition (by Analyte or Components) Pd 26.3% min.
Odor Odorless
Sensitivity Hygroscopic
Density 2.738 g/mL
Chemical Name or Material Potassium hexachloropalladate(IV)
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.