Learn More
Alfa Aesar™ Potassium hexachloropalladate(IV), Pd 26.3% min
Brand: Alfa Aesar™ 011884.06
Code : NEW
Additional Details : CAS Number : 16919-73-6 Weight : 0.00500kg Transport : UN number : 3077 Chem class : 1 Pack group : III
Quantity | 5g |
---|
Chemical Identifiers
Potassium hexachloropalladate(IV) | |
397.32 | |
UN3077 |
16919-73-6 | |
MFCD00043081 |
Specifications
Potassium hexachloropalladate(IV) | |
Crystalline | |
MFCD00043081 | |
Pd 26.3% min. | |
Odorless | |
Red |
16919-73-6 | |
397.32 | |
UN3077 | |
2.738g/mL | |
Hygroscopic | |
5g |