missing translation for 'onlineSavingsMsg'
Learn More

Potassium hexachloroosmate(IV), Os 38.7% min, Thermo Scientific Chemicals

Product Code. 11306568
Click to view available options
Quantity:
0.25 g
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Potassium hexachloroosmate(IV) used as Pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16871-60-6
MDL Number MFCD00011371

Specifications

Melting Point (decomposition)
Color Red
Physical Form Powder
Quantity 0.25 g
Linear Formula K2OsCl6
Merck Index 14,7635
Solubility Information Soluble in water; Sparingly soluble in alcohol
Formula Weight 481.12
Concentration or Composition (by Analyte or Components) Os 38.7% min.
Sensitivity Hygroscopic
Chemical Name or Material Potassium hexachloroosmate(IV)
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'