missing translation for 'onlineSavingsMsg'
Learn More

Potassium hexachloroosmate(IV), min 39% Os

Product Code. 10358670 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10358670 5 g Glass bottle 5g
10348670 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10358670 Supplier Thermo Scientific Acros Supplier No. 197650050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 16871-60-6 | Cl6K2Os | 481.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16871-60-6
Molecular Formula Cl6K2Os
Molecular Weight (g/mol) 481.11
MDL Number MFCD00011371

Specifications

Melting Point 600.0°C
Color Bordeaux to Purple
Physical Form Powder
Quantity 5 g
Linear Formula K2OsCl6
Merck Index 15, 7756
Solubility Information Solubility in water: soluble. Other solubilities: sparingly soluble in alcohol
Formula Weight 481.11
Percent Purity ≥39% (Os)
Packaging Glass bottle
Density 3.42
Chemical Name or Material Potassium hexachloroosmate(IV), min 39% Os
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Potassium hexachloroosmate(IV)
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.