missing translation for 'onlineSavingsMsg'
Learn More

Potassium hexachloroiridate(III) hydrate, Ir 32.8% min

Product Code. 11361978
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11361978 5 g 5g
11341978 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11361978 Supplier Thermo Scientific Alfa Aesar Supplier No. 011887.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 35705-96-5

Potassium hexachloroiridate(III) hydrate is used to prepare other iridium compounds. It is used as a redox mediator to visualize human finger prints on a poly(vinylidene difluoride) membrane by scanning electrochemical microscopy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35705-96-5
MDL Number MFCD00038699

Specifications

Physical Form Crystalline Powder
Quantity 5 g
Solubility Information Soluble in water.
Formula Weight 522.22 (Anhydrous)
Concentration or Composition (by Analyte or Components) Ir 33.4% min.
Chemical Name or Material Potassium hexachloroiridate(III) hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.